Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STING/TMEM173 Mouse anti-Human, Clone: STING1/7432, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP313942100UG
This item is not returnable.
View return policy
Description
STING/TMEM173 Monoclonal antibody specifically detects STING/TMEM173 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| STING/TMEM173 | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 ug/ml | |
| Q86WV6 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| STING1/7432 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| endoplasmic reticulum IFN stimulator, Endoplasmic reticulum interferon stimulator, ERIS, FLJ38577, hMITA, hSTING, Mediator of IRF3 activation, MITA, mitochondrial mediator of IRF3 activation, MPYS, NET23, N-terminal methionine-proline-tyrosine-serine plasma membrane tetraspanner, SAVI, Stimulator of interferon genes protein, stimulator of interferon protein, sting, STING-beta, TMEM173, transmembrane protein 173 | |
| Recombinant fragment (around aa190-290) of human STING/TMEM173 protein (exact sequence is proprietary) (Uniprot: Q86WV6 ) | |
| 100 μg | |
| Cancer, Immunology, Innate Immunity | |
| 340061 | |
| Store at 4°C. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction