Learn More
Invitrogen™ STIP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589626
Description
Immunogen sequence: MEQVNELKEK GNKALSVGNI DDALQCYSEA IKLDPHNHVL YSNRSAAYAK KGDYQKAYED GCKTVDLKPD WGKGYSRKAA ALEFLNRFEE AKRTYEEGLK HEANNPQLKE GLQNMEARLA ERKFMNPFNM PNLYQKLESD PRTRTLLSDP TYRELIEQLR NKPSDLGTKL QDPRIMTTLS VLLGVDLGSM DEEEEIATPP PPPPPKKETK PEPMEEDLPE NKKQALKEKE LGNDAYKKKD FDTALKHYDK AKELDPTNMT YITNQAAVYF EKGDYNKCRE LCEKAIEVGR ENREDYRQIA; Positive Samples: HeLa, COS-7, COS-1, OVCAR3, Mouse brain; Cellular Location: Cytoplasm, Nucleus.
Mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).
Specifications
STIP1 | |
Polyclonal | |
Unconjugated | |
STIP1 | |
epididymis secretory sperm binding protein Li 94n; HEL-S-94n; HOP; hsc70/Hsp90-organizing protein; Hsp70/Hsp90 organizing protein; Hsp70/Hsp90-organizing protein; IEF SSP 3521; IEF-SSP-3521; mSTI1; NY-REN-11 antigen; P60; Renal carcinoma antigen NY-REN-11; STI1; STI1L; Stip1; stress induced phosphoprotein 1; stress-induced phosphoprotein 1; stress-induced-phosphoprotein 1; stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein); stress-inducible protein; transformation-sensitive protein IEF SSP 3521 | |
Rabbit | |
Affinity chromatography | |
RUO | |
10963, 192277, 20867 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35814, P31948, Q60864 | |
STIP1 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human STIP1 (NP_0068101). | |
100 μL | |
Primary | |
Human, Mouse, Rat, Monkey | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.