Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STIP1 Recombinant Rabbit Monoclonal Antibody (3E4)
GREENER_CHOICE

Catalog No. PIMA533200
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533200 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533200 Supplier Invitrogen™ Supplier No. MA533200
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

STIP1 Recombinant Monoclonal Antibody for Western Blot, ELISA

Mediates the association of the molecular chaperones HSC70 and HSP90 (HSPCA and HSPCB).
TRUSTED_SUSTAINABILITY

Specifications

Antigen STIP1
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 3E4
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene STIP1
Gene Accession No. P31948
Gene Alias epididymis secretory sperm binding protein Li 94n; HEL-S-94n; HOP; hsc70/Hsp90-organizing protein; Hsp70/Hsp90 organizing protein; Hsp70/Hsp90-organizing protein; IEF SSP 3521; IEF-SSP-3521; mSTI1; NY-REN-11 antigen; P60; Renal carcinoma antigen NY-REN-11; STI1; STI1L; Stip1; stress induced phosphoprotein 1; stress-induced phosphoprotein 1; stress-induced-phosphoprotein 1; stress-induced-phosphoprotein 1 (Hsp70/Hsp90-organizing protein); stress-inducible protein; transformation-sensitive protein IEF SSP 3521
Gene Symbols STIP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human STIP1.
Purification Method Antigen Affinity Chromatography, Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10963
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.