Learn More
STK11 Mouse anti-Human, Clone: OTI0A3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF802554

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
LKB1 (also known as STK11 - Serine/threonine-protein kinase STK11), a member of the serine/threonine kinase family, is a tumor suppressor. LKB1 physically associates with p53 and regulates specific p53 dependent apoptosis pathways. LKB1 is present in both the cytoplasm and nucleus of living cells and translocates to mitochondria during apoptosis. LKB1 functions as a master upstream protein kinase, regulating AMPK related kinases as well as AMPK. Mutations in LKB1 gene are associated with Peutz Jeghers syndrome.Specifications
STK11 | |
Monoclonal | |
Unconjugated | |
STK11 | |
LKB1, PJS, hLKB1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
6794 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI0A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15831 | |
STK11 | |
Human recombit protein fragment corresponding to amino acids 1-236 of human STK11 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.