Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK11IP Antibody, Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB261925 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB261925 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB261925 Supplier Novus Biologicals Supplier No. NBP130110AF532
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STK11IP Polyclonal antibody specifically detects STK11IP in Human samples. It is validated for Western Blot

Specifications

Antigen STK11IP
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 532
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias KIAA1898LKB1 interacting protein, LIP1serine/threonine kinase 11-interacting protein, LKB1-interacting protein 1, LKB1IPserine/threonine-protein kinase 11-interacting protein, serine/threonine kinase 11 interacting protein, STK11IP1STK11 interacting protein
Host Species Rabbit
Immunogen maps to a region between residue 900 and 950 of human serine/threonine kinase 11 interacting protein using the numbering given in entry NP_443134.2
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114790
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.