Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK11IP Antibody, DyLight 755, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP130110IR
Description
STK11IP Polyclonal antibody specifically detects STK11IP in Human samples. It is validated for Western BlotSpecifications
STK11IP | |
Polyclonal | |
Western Blot | |
KIAA1898LKB1 interacting protein, LIP1serine/threonine kinase 11-interacting protein, LKB1-interacting protein 1, LKB1IPserine/threonine-protein kinase 11-interacting protein, serine/threonine kinase 11 interacting protein, STK11IP1STK11 interacting protein | |
maps to a region between residue 900 and 950 of human serine/threonine kinase 11 interacting protein using the numbering given in entry NP_443134.2 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 755 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
114790 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction