Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK11IP Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982411
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2411 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2411 Supplier Bioss Supplier No. BS12827R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

May regulate STK11/LKB1 function by controlling its subcellular localization.

Specifications

Antigen STK11IP
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene STK11IP
Gene Accession No. Q8N1F8
Gene Alias KIAA1898; LIP1; LKB1-interacting protein 1; LKB1IP; serine/threonine kinase 11 interacting protein; Serine/threonine-protein kinase 11-interacting protein; STK11 interacting protein; STK11IP; STK11IP1
Gene Symbols STK11IP
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human STK11IP.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114790
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.