Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566998
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566998 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566998 Supplier Thermo Scientific Supplier No. PA566998
Add to Cart
Add to Cart

STK19 Polyclonal Antibody

STK19 Polyclonal Antibody for ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen STK19
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STK19
Gene Accession No. P49842
Gene Alias D6S60; D6S60E; G11; HLA-RP1; MHC class III HLA-RP1; nuclear serine/threonine protein kinase; Protein G11; Protein RP1; RP1; serine/threonine kinase 19; serine/threonine-protein kinase 19; Stk19
Gene Symbols STK19
Host Species Rabbit
Immunogen Recombinant Human STK19. Recombinant protein control fragment (Product #RP-107640).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8859
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.