Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STK19 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116560
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116560 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116560 Supplier Invitrogen™ Supplier No. PA5116560
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: Mouse testis, mouse liver, rat lung Immunogen sequence: GTPGETVRHC SAPEDPIFRF SSLHSYPFPG TIKSRDMSWK RHHLIPETFG VKRRRKRGPV ESDPLRGEPG SARAAVSELM QLFPRGLFED ALPPIVLRSQ VYSLVPDRTV ADRQLKELQE QGEIRIVQLG FDLDAHGIIF TEDYRTRVLK ACDGRPYAGA VQKFLASVLP ACGDLSFQQD QMTQTFGFRD SEITHLVNAG VLTVRDAGSW WLAVPGAGRF IKYFVKGRQA VLSMVRKAKY RELLLSELLG RRAPVVVRLG LTYHVHDLIG AQLVDCISTT SGTLLRLPET.

This gene encodes a serine/threonine kinase which localizes predominantly to the nucleus. Its specific function is unknown; it is possible that phosphorylation of this protein is involved in transcriptional regulation. This gene localizes to the major histocompatibility complex (MHC) class III region on chromosome 6 and expresses two transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK19
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.69 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STK19
Gene Accession No. P49842, Q9JHN8
Gene Alias D6S60; D6S60E; G11; HLA-RP1; MHC class III HLA-RP1; nuclear serine/threonine protein kinase; Protein G11; Protein RP1; RP1; serine/threonine kinase 19; serine/threonine-protein kinase 19; Stk19
Gene Symbols STK19
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 75-364 of human STK19 (NP_0041881).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361800, 54402, 8859
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.