Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human STK22D (TSSK1), His Tag Recombinant Protein

Catalog No. PV3505
Encompass_Preferred
Click to view available options
Quantity:
10 μg

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Protein Form: Recombinant, Full Length

STK22D (TSSK1) is a serine/threonine kinase essential to the function and production of sperm (spermiogenesis). TSSK1 belongs to a family of serine/threonine kinases highly expressed in testis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9BXA7
Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.05% NP-40, 0.5 mM EDTA, 100 mM NaCl, 4 mM DTT, 50% glycerol and no preservative; pH 7.5
Gene ID (Entrez) 83942
Molecular Weight (g/mol) 46.7 kDa
Name Human STK22D (TSSK1), His Tag
Quantity 10 μg
Regulatory Status For research use only. Not for use in diagnostic procedures
Gene Alias FKSG81; serine/threonine kinase 22 A (spermiogenesis associated); serine/threonine kinase 22 D (spermiogenesis associated); serine/threonine kinase FKSG81; serine/threonine-protein kinase 22 A; spermiogenesis associated; spermiogenesis associated 4; SPOGA1; SPOGA4; Stk22a; STK22D; testis specific serine kinase 1 B; testis tissue sperm-binding protein Li 55 e; testis-specific kinase 1; testis-specific serine kinase 1; testis-specific serine kinase 1 B; testis-specific serine/threonine kinase; testis-specific serine/threonine-protein kinase 1; TSK1; TSK-1; Tssk; TSSK1; TSSK-1; TSSK1B
Common Name TSSK1
KinaseFamily TSSK Kinase family
Product Type STK22D (TSSK1) Recombinant Human Protein
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-tag
Sequence Full length
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Insect cells
Protein Family Kinases and inhibitors
Form Liquid
Protein Subtype Serine/threonine kinase
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.