Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STK32A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559115
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559115 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559115 Supplier Invitrogen™ Supplier No. PA559115
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HIRSSTSSKE IVHTFETTVV TYPSAWSQEM VSLLKKLLEP NPDQRFSQLS DVQNFPYMND INWDAVFQKR LIPGFIP Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 84%.

STK32A (serine/threonine kinase 32A), also known as YANK1, is a 396 amino acid protein that belongs to the superfamily of serine/threonine protein kinases and exists as three isoforms. The gene encoding STK32A maps to human chromosome 5, which is associated with Cockayne syndrome through the ERCC8 gene and familial adenomatous polyposis through the adenomatous polyposis coli (APC) tumor suppressor gene. Treacher Collins syndrome is also chromosome 5 associated and is caused by insertions or deletions within the TCOF1 gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK32A
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STK32A
Gene Accession No. Q8WU08
Gene Alias A930015B13Rik; CTB-108O6.2; serine/threonine kinase 32A; serine/threonine-protein kinase 32A; Stk32a; YANK1; Yet another novel kinase 1
Gene Symbols STK32A
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STK32A. Recombinant protein control fragment (Product #RP-97248).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 202374
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.