Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK32A Polyclonal Antibody
STK32A Polyclonal Antibody
Supplier: Thermo Scientific PA570134
Description
STK32A Polyclonal Antibody for Western Blot

Specifications
STK32A | |
Polyclonal | |
Unconjugated | |
STK32A | |
A930015B13Rik; CTB-108O6.2; serine/threonine kinase 32A; serine/threonine-protein kinase 32A; Stk32a; YANK1; Yet another novel kinase 1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
202374 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8WU08 | |
STK32A | |
synthetic peptide directed towards the N-terminal region of human STK32A. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction