Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STK32B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566178
Description
Immunogen sequence: LRKLLTKDPES RVSSLHDIQS VPYLADMNWD AVFKKALMPG FV Highest antigen sequence identity to the following orthologs - mouse 86%, rat 84%.
This gene encodes a serine-threonine protein kinase. Serine-threonine kinases transfer phosphate molecules to the oxygen atoms of serine and threonine. A genomic deletion affecting this gene has been associated with Ellis-van Creveld syndrome, an autosomal recessive skeletal dysplasia. Alternative splicing results in multiple transcript variants.
Specifications
STK32B | |
Polyclonal | |
Unconjugated | |
STK32B | |
2510009F08Rik; gene for serine/threonine protein kinase; HSA250839; serine threonine kinase 32; serine/threonine kinase 32B; serine/threonine-protein kinase 32B; Stk32; Stk32b; STKG6; UNQ3003/PRO9744; YANK2; Yet another novel kinase 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55351 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NY57 | |
STK32B | |
Recombinant Human STK32B. Recombinant protein control fragment (Product #RP-106866). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction