Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human STK32B (YANK2), GST Tag Recombinant Protein

Catalog No. PV6406
Encompass_Preferred
Click to view available options
Quantity:
1 mg
10 μg
100 μg

Recombinant Protein

For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Full-length recombinant human STK32B (YANK2) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. , Validated in LanthaScreen™ Kinase Binding Assay, Protocols available for simple use and optimized performance, Multiple pack sizes available for convenient use Related Product: LanthaScreen™ Eu Kinase Binding Assay Protein Form: Recombinant

STK32B or YANK2 is a serine/threonine protein kinase that contains motif for binding metal ions and nucleotides and the gene for STK32B is highly conserved among various species. STK32B has been associated with isolated cleft lip with or without cleft palate and cleft palate which are among the most common human birth defects of oral cleft cases. Long interspersed nuclear element-1 (LINE-1 or L1) mediated deletion of STK32B gene is observed in patients with Ellis-van Creveld syndrome with borderline intelligence.
TRUSTED_SUSTAINABILITY

Specifications

Concentration See Label
For Use With (Application) Kinase Assay
Formulation 50 mM tris HCl with 0.1 mM EDTA, 0.1 mM PMSF, 0.25 mM DTT, 10 mM glutathione, 150 mM NaCl, 25% glycerol and no preservative; pH 7.5
Molecular Weight (g/mol) 74.1 kDa
Name Human STK32B (YANK2), GST Tag
Quantity 10 μg
Common Name STK32B (R198G Mutant)
Protein Length 1-414 [R198G]
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag GST-tag
Content And Storage -80°C, Avoid Freeze/Thaw Cycles
Expression System Baculovirus
Form Liquid
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.