Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STK32C Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529414
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529414 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529414 Supplier Invitrogen™ Supplier No. PA529414
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431, Molt-4, Raji, mouse brain. Predicted reactivity: Mouse (96%), Rat (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

STK32C or YANK3 is a member of the YANK family and is a serine/threonine protein kinase of approximately 486 aa that requires magnesium for catalytic activity. There are two isoforms of the protein generated by alternative splicing and the protein undergoes serine phosphorylation by upstream protein kinases. The STK32C gene product is conserved in many species including chimpanzee, dog, cow, mouse, rat, and chicken.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK32C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.43 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene Stk32c
Gene Accession No. Q86UX6, Q8QZV4
Gene Alias MNCb-1563; PKE; PKE protein kinase; Pkek; RP11-140A10.1; serine/threonine kinase 32C; serine/threonine-protein kinase 32C; STK32C; testicular tissue protein Li 187; YANK3; Yet another novel kinase 3
Gene Symbols Stk32c
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human STK32C. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 282974, 57740
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.