Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STK32C Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553662
Description
Immunogen sequence: PTFELEEMIL ESRPLHKKKK RLAKNKSRDN SRDSSQSEND YLQDCLDAIQ QDFVIFNREK LKRSQDLPRE PLPAPESRDA AEPVEDEAER SALPMCGPIC PS Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
STK32C or YANK3 is a member of the YANK family and is a serine/threonine protein kinase of approximately 486 aa that requires magnesium for catalytic activity. There are two isoforms of the protein generated by alternative splicing and the protein undergoes serine phosphorylation by upstream protein kinases. The STK32C gene product is conserved in many species including chimpanzee, dog, cow, mouse, rat, and chicken.
Specifications
STK32C | |
Polyclonal | |
Unconjugated | |
Stk32c | |
MNCb-1563; PKE; PKE protein kinase; Pkek; RP11-140A10.1; serine/threonine kinase 32C; serine/threonine-protein kinase 32C; STK32C; testicular tissue protein Li 187; YANK3; Yet another novel kinase 3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
282974 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q86UX6 | |
Stk32c | |
Recombinant protein corresponding to Human STK32C. Recombinant protein control fragment (Product #RP-91474). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction