Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK32C Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 114341AP150UL

Description
The protein encoded by this gene is a member of the serine/threonine protein kinase family. The specific function of this kinase is not known.Specifications
STK32C | |
Polyclonal | |
Unconjugated | |
STK32C | |
PKE, RP11 140A10.1, serine/threonine kinase 32C, STK32C, YANK3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
282974, 365381, 57740 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q86UX6, Q8QZV4 | |
STK32C | |
STK32C Fusion Protein Ag1960 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction