Learn More
Promega STK33 Kinase Enzyme System, 10microg
Catalog No. PRV4086 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV4086 10 μg
1 options
Catalog No. PRV4086 Supplier Promega Supplier No. V4086
Recombinant full-length human STK33 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. STK33 is a distant member of the CAMK group of serine/threonine kinases.
  • Easily Screen and Profile STK33 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C