Learn More
Promega STK33 Kinase Enzyme System, 1mg
Catalog No. PRV6563 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
1 mg
Catalog No. Quantity
PRV6563 1 mg
1 options
Catalog No. PRV6563 Supplier Promega Supplier No. V6563
Add to Cart
Add to Cart
Request a Quote
Recombinant full-length human STK33 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. STK33 is a distant member of the CAMK group of serine/threonine kinases.
  • Easily Screen and Profile STK33 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 1 mg
Content And Storage <-65°C