Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK33 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982186
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2186 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2186 Supplier Bioss Supplier No. BS12373R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STK33 gene ontology annotations related to this gene include ATP binding; protein kinase activity.

Specifications

Antigen STK33
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene STK33
Gene Accession No. Q9BYT3
Gene Alias 4921505G21Rik; AW061264; serine/threonine kinase 33; Serine/threonine-protein kinase 33; Stk33
Gene Symbols STK33
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human STK33.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65975
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.