Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STK35 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110441
Description
Immunogen sequence: RPEGGGGSAR PRYSLLAEIG RGSYGVVYEA VAGRSGARVA VKKIRCDAPE NVELALAEFW ALTSLKRRHQ NVVQFEECVL QRNGLAQRMS HGNKSSQLYL RLVETSLKGE RILGYAEEPC YLWFVMEFCE GGDLNQYVLS RRPDPATNKS FMLQLTSAIA FLHKNHIVHR DLKPDNILIT ERSGTPILKV A.
The protein encoded by this gene is a kinase that is predominantly found in the nucleus. However, it can interact with PDLIM1/CLP-36 in the cytoplasm and localize to actin stress fibers. The encoded protein may be a regulator of actin stress fibers in nonmuscle cells.
Specifications
STK35 | |
Polyclonal | |
Unconjugated | |
Stk35 | |
1700054C12Rik; bA550O8.2; bA550O8.2 CLIK1; CLIK1; CLIK-1; CLP-36 interacting kinase; CLP-36-interacting kinase 1; OTTHUMP00000030006; PDIK1; PDLIM1-interacting kinase 1; serine threonine kinase 35 long form; serine/threonine kinase 35; serine/threonine-protein kinase 35; Serine/threonine-protein kinase 35 L1; STK35; Stk35l1 | |
Rabbit | |
Affinity chromatography | |
RUO | |
67333 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.89 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q80ZW0 | |
Stk35 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 190-380 of human STK35 (NP_543026.2). | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction