Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK35 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982413
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2413 Supplier Bioss Supplier No. BS12834R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a kinase that is predominantly found in the nucleus. However, it can interact with PDLIM1/CLP-36 in the cytoplasm and localize to actin stress fibers. The encoded protein may be a regulator of actin stress fibers in nonmuscle cells.

Specifications

Antigen STK35
Applications Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Stk35
Gene Accession No. Q8TDR2
Gene Alias 1700054C12Rik; bA550O8.2; bA550O8.2 CLIK1; CLIK1; CLIK-1; CLP-36 interacting kinase; CLP-36-interacting kinase 1; OTTHUMP00000030006; PDIK1; PDLIM1-interacting kinase 1; serine threonine kinase 35 long form; serine/threonine kinase 35; Serine/threonine-protein kinase 35; Serine/threonine-protein kinase 35 L1; STK35; STK35L1
Gene Symbols Stk35
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human STK35.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140901
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.