Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STK36 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555603
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555603 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555603 Supplier Invitrogen™ Supplier No. PA555603
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LLYHPFIAGH VTIITEPAGP DLGTPFTSRL PPELQVLKDE QAHRLAPKGN QSRILTQAYK RMAEEAMQKK HQNTGPALEQ EDKTSKVAPG T Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 81%.

Serine/threonine protein kinase required for postnatal development, possibly by regulating the homeostasis of cerebral spinal fluid or ciliary function. Controls the activity of the transcriptional regulators GLI1, GLI2 and GLI3 by opposing the effect of SUFU and promoting their nuclear localization. GLI2 requires an additional function of STK36 to become transcriptionally active, but the enzyme does not need to possess an active kinase catalytic site for this to occur.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK36
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STK36
Gene Accession No. Q9NRP7
Gene Alias 1700112N14Rik; B930045J24; FU; Fused; Fused homolog; Kiaa1278; mKIAA1278; serine/threonine kinase 36; serine/threonine kinase 36 (fused homolog, Drosophila); serine/threonine-protein kinase 36; STK36; testicular tissue protein Li 188
Gene Symbols STK36
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STK36. Recombinant protein control fragment (Product #RP-95075).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27148
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.