Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STK38 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
SDP

Catalog No. 501736115 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6115 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6115 Supplier Proteintech Group Inc Supplier No. 553351AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody recognizes the C-terminal of STK38.

Stk38,also known as NUCLEAR DBF2-RELATED PROTEIN, contains 1 protein kinase domain that interacts with mob1 and mob2.The homodimeric s100b binds to two molecules of stk38. It is ubiquitously expressed with highest levels observed in peripheral blood leukocytes. Stk38 activated by binding of s100b which releases autoinhibitory n-lobe interactions, enabling atp to bind and the autophosphorylation of ser-281. Thr-444 then undergoes calcium- dependent phosphorylation by an upstream kinase. Interactions between phosphorylated Thr-444 and the n-lobe promote additional structural changes that complete the activation of the kinase. Autoinhibition is also released by the binding of mob1/mobkl1a and mob2/hcca2 to the n-terminal of stk38.

Specifications

Antigen STK38
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STK38
Gene Accession No. Q15208, Q91VJ4
Gene Alias NDR, NDR1, NDR1 protein kinase, Nuclear Dbf2 related kinase 1, serine/threonine kinase 38, STK38
Gene Symbols Stk38
Host Species Rabbit
Immunogen Peptide
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 106504, 11329
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.