Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK38L Antibody (OTI4D8), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274403R
Description
STK38L Monoclonal specifically detects STK38L in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
STK38L | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
STK38L | |
Recombinant protein fragment corresponding to amino acids 1-281 of human STK38L (NP_055815) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat, Canine, Monkey | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI4D8 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
EC 2.7.11, EC 2.7.11.1, KIAA0965nuclear Dbf2-related 2, NDR2 protein kinase, NDR2serine/threonine-protein kinase 38-like, Nuclear Dbf2-related kinase 2, serine/threonine kinase 38 like | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
23012.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction