Learn More
STK38L Mouse anti-Human, Clone: OTI3E10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF505210

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
STK38L contains 12 protein kinase catalytic subdomains and a potential bipartite nuclear localization signal. Northern blot analysis revealed expression of a 3.9-kb transcript in all tissues tested, with the possible exception of adult brain. Highest expression was in peripheral blood leukocytes. Immunoblot and immunofluorescence microscopy demonstrated predomitly nuclear expression of a 55-kD protein.Specifications
STK38L | |
Monoclonal | |
Unconjugated | |
STK38L | |
NDR2 | |
Mouse | |
Affinity Chromatography | |
RUO | |
23012 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3E10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q9Y2H1 | |
STK38L | |
Human recombit protein fragment corresponding to amino acids 1-281 of human STK38L produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.