Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STK38L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120359
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120359 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120359 Supplier Invitrogen™ Supplier No. PA5120359
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: HeLa, Mouse small intestine, Mouse brain, Rat stomach, Rat brain. The target is usually found in the following locations: Cytoplasm, Membrane, cytoskeleton. Immunogen sequence: LIMEFLPGGD MMTLLMKKDT LTEEETQFYI SETVLAIDAI HQLGFIHRDI KPDNLLLDAK GHVKLSDFGL CTGLKKAHRT EFYRNLTHNP PSDFSFQNMN SKRKAETWKK NRRQLAYSTV GTPDYIAPEV FMQTGYNKLC DWWSLGVIMY EMLIGYPPFC SETPQETYRK VMNWKETLVF PPEVPISEKA KDLILRFCID SENRIGNSGV EEIKGHPFFE GVDWEHIRER PAAIPIEIKS IDDTSNFDDF PESDILQPVP NTTEPDYKSK DWVFLNYTYK RFEGLTQRGS IPTYMKAGKL.

STK38L (also known as NDR2) or Nuclear Dbf2-related protein is a serine/threonine kinase in the subgroup of the AGC kinase family. STK38L act as novel pro-apoptotic kinase that is activated by RASSF1A/MST1 in response to Fas receptor stimulation and promotes apoptosis. STK38L is mainly involved in the regulation of structural processes in differentiating and mature neuronal cells and it is highly expressed with highest levels observed in the thymus. STK38L contains 12 protein kinase catalytic subdomains and a potential bipartite nuclear localization signal.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STK38L
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Stk38l
Gene Accession No. Q7TSE6, Q9Y2H1
Gene Alias 4930473A22Rik; B230328I19; KIAA0965; Ndr2; NDR2 protein kinase; Ndr54; nuclear Dbf2-related 2; nuclear Dbf2-related kinase 2; RGD1564793; serine/threonine kinase 38 like; serine/threonine kinase 38-like; serine/threonine-protein kinase 38-like; STK38L
Gene Symbols Stk38l
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 165-464 of human STK38L (NP_055815.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23012, 232533, 691337
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.