Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK39 Antibody (OTI4E3), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274406PE
Description
STK39 Monoclonal specifically detects STK39 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.Specifications
STK39 | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
STK39 | |
Full length recombinant protein of human STK39 (NP_037365) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Flow Cytometry | |
OTI4E3 | |
Flow Cytometry | |
DCHTsmall intestine SPAK-like kinase, DKFZp686K05124, EC 2.7.11, EC 2.7.11.1, PASK, proline-alanine-rich STE20-related kinase, serine threonine kinase 39, Serine/threonine-protein kinase 39, SPAKserine threonine kinase 39 (STE20/SPS1 homolog, yeast), STE20/SPS1 homolog, STE20/SPS1-related proline-alanine-rich protein kinase, Ste20-like protein kinase, Ste-20-related kinase | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
27347.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction