Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STK39 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50 - $596.50
Specifications
Antigen | STK39 |
---|---|
Concentration | 0.05 mg/mL |
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111538
|
Invitrogen™
PA5111538 |
100 μL |
Each for $596.50
|
|
|||||
PIPA5111553
|
Invitrogen™
PA5111553 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: IVDDPKALKT LTFKLASGCD GSEIPDEVKL.
STK39 (STLK3) is involved in the cellular stress response pathway. STK39 is activated in response to hypotonic stress leading to phosphorylation of several cation-chloride-coupled co-transporters. STK39 activates the p38 MAP kinase pathway and its interaction with p38 decreases during cellular stress. STK39 acts as an intermediate in the response to cellular stress. STK39 is also an independent risk factor for hypertension in men and its intragenic SNPs can interact and function in the control of blood pressure.Specifications
STK39 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
STK39 | |
AW227544; AW556857; DCHT; DKFZp686K05124; kidney-specific SPAK; pancreatic serine/threonine-protein kinase; Pask; proline-alanine-rich STE20-related kinase; PS/TK; PSTK1; RF005; Rnl5; serine threonine kinase 39; serine threonine kinase 39 (STE20/SPS1 homolog, yeast); serine/threonine kinase 39; serine/threonine kinase 39, STE20/SPS1 homolog; serine/threonine-protein kinase 39; small intestine SPAK-like kinase; Spak; Ste-20 related kinase; STE20/SPS1 homolog; STE20/SPS1-related proline-alanine-rich protein kinase; Ste20-like protein kinase; ste-20-related kinase; Stk39 | |
STK39 | |
Primary | |
Antigen affinity chromatography |
0.05 mg/mL | |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UEW8 | |
27347 | |
Recombinant Protein corresonding to Human STK39. Recombinant protein control fragment (Product #RP-104045). | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title