Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK4 Antibody, DyLight 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11039128G
Description
STK4 Polyclonal antibody specifically detects STK4 in Human, Mouse samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
STK4 | |
Polyclonal | |
Western Blot, Immunohistochemistry | |
EC 2.7.11, EC 2.7.11.1, kinase responsive to stress 2, KRS2KRS2)), Mammalian STE20-like protein kinase 1, mammalian sterile 20-like 1, MST-1, MST1DKFZp686A2068, serine/threonine kinase 4, serine/threonine-protein kinase 4, Serine/threonine-protein kinase Krs-2, STE20-like kinase MST1 | |
The immunogen recognized by this antibody maps to the C-terminus of human Mammalian Sterile 20-like 1 (Serine/Threonine Kinase 4) using the numbering given in Swiss Prot entry Q13043 (GeneID 6789). | |
0.1 mL | |
Apoptosis, Phospho Specific | |
6789 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunohistochemistry | |
DyLight 488 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction