Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STK4 Antibody, PerCP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299813PCP
Description
STK4 Polyclonal antibody specifically detects STK4 in Human samples. It is validated for ELISASpecifications
STK4 | |
Polyclonal | |
ELISA | |
EC 2.7.11, EC 2.7.11.1, kinase responsive to stress 2, KRS2KRS2)), Mammalian STE20-like protein kinase 1, mammalian sterile 20-like 1, MST-1, MST1DKFZp686A2068, serine/threonine kinase 4, serine/threonine-protein kinase 4, Serine/threonine-protein kinase Krs-2, STE20-like kinase MST1 | |
Produced in rabbits immunized with purified, recombinant Human STK4 (Uniprot#: Q13043-1; Glu2-Phe487) | |
0.1 mL | |
Apoptosis, Phospho Specific | |
6789 | |
Store at 4C in the dark. | |
IgG |
ELISA | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction