Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STKLD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556290
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556290 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556290 Supplier Invitrogen™ Supplier No. PA556290
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CQDRALLVNN AYRGLASLVK VSELAAFKVV VQEEGGSGLS LIKETYQLHR DDPEVVENVG MLLVHLASYE EILPELVSSS MKALLQEIKE RFTSSLVSDS S Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.

STKLD1 (Serine/Threonine Kinase Like Domain Containing 1) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene includetransferase activity, transferring phosphorus-containing groupsandprotein tyrosine kinase activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STKLD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STKLD1
Gene Accession No. Q8NE28
Gene Alias C9orf96; Gm711; likely ortholog of H. sapiens chromosome 9 open reading frame 96 (C9orf96); LOC169436; probable inactive protein kinase-like protein SgK071; probable inactive protein kinase-like protein SgK071 homolog; probable protein kinase-like protein SgK071 homolog; RP11-244N20.8; serine/threonine kinase like domain containing 1; serine/threonine kinase-like domain containing 1; serine/threonine kinase-like domain-containing protein 1; Serine/threonine kinase-like domain-containing protein STKLD1; SGK071; Sk521; STKLD1; Sugen kinase 071
Gene Symbols STKLD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STKLD1. Recombinant protein control fragment (Product #RP-95247).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 169436
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.