Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STMN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518034
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518034 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518034 Supplier Invitrogen™ Supplier No. PA518034
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

STMN2, also known as SCG-10, is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin. STMN2 may play a role in neuronal differentiation, and in modulating membrane interaction with the cytoskeleton during neurite outgrowth. Reductions in the expression of this gene have been associated with Down's syndrome and Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STMN2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Stmn2
Gene Accession No. P55821
Gene Alias AI159727; hypothetical protein LOC534991; neuronal growth-associated protein (silencer element); neuron-specific growth-associated protein; Protein SCG10; SCG10; Scgn10; SGC10; stathmin 2; stathmin-2; stathmin-like 2; Stmb2; Stmn2; superior cervical ganglia, neural specific 10; superior cervical ganglion-10 protein; superiorcervical ganglia, neural specific 10
Gene Symbols Stmn2
Host Species Goat
Immunogen Synthetic peptide sequence (AKTAMAYKEK) corresponding to the N-terminus amino acids of STMN2 (aa 2-12).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20257
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.