Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STMN2/SCG10 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985627
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5627 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5627 Supplier Bioss Supplier No. BS3753R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

STMN2, also known as SCG-10, is a neuronal growth-associated protein that shares significant amino acid sequence similarity with the phosphoprotein stathmin. STMN2 may play a role in neuronal differentiation, and in modulating membrane interaction with the cytoskeleton during neurite outgrowth. Reductions in the expression of this gene have been associated with Down′s syndrome and Alzheimer′s disease.

Specifications

Antigen STMN2/SCG10
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Stmn2
Gene Accession No. Q93045
Gene Alias AI159727; BOS_13924; neuronal growth-associated protein (silencer element); neuron-specific growth-associated protein; Protein SCG10; SCG10; Scgn10; SGC10; stathmin 2; Stathmin-2; stathmin-like 2; Stmb2; Stmn2; superior cervical ganglia, neural specific 10; Superior cervical ganglion-10 protein; superiorcervical ganglia, neural specific 10
Gene Symbols Stmn2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human STMN2/SCG10.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11075
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.