Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
STMN3 Mouse anti-Human, Clone: 1F12B1, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | STMN3 |
---|---|
Clone | 1F12B1 |
Concentration | 0.55 mg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6586
![]() ![]() |
Proteintech Group Inc
661521IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
This gene encodes a protein which is a member of the stathmin protein family. Members of this protein family form a complex with tubulins at a ratio of 2 tubulins for each stathmin protein. Microtubules require the ordered assembly of alpha- and beta-tubulins, and formation of a complex with stathmin disrupts microtubule formation and function. A pseudogene of this gene is located on chromosome 22. Alternative splicing results in multiple transcript variants.Specifications
STMN3 | |
0.55 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
SCG10 like protein, SCLIP, Stathmin 3, stathmin like 3, STMN3 | |
STMN3 | |
IgG2b | |
Protein G | |
Antibody |
1F12B1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9NZ72 | |
50861 | |
STMN3 Fusion Protein Ag18844 | |
Primary | |
-20°C | |
STMN3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title