Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STMN4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101915
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101915 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101915 Supplier Invitrogen™ Supplier No. PA5101915
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total STMN4.

RB3 (Stathmin-4), encoded by STMN4 gene, is a neuron-specific homologue of the SCG10/stathmin family proteins, possessing a unique N-terminal membrane-associated domain and the stathmin-like domain at the C terminus, which promotes microtubule (MT) catastrophe and/or tubulin sequestering. Stathmin is a ubiquitous cytosolic phosphoprotein, preferentially expressed in the nervous system, and described as a relay integrating diverse intracellular signaling pathways. RB3 is consistently up-regulated in differentiated neuroblastoma cells, and stable expression of transfected RB3 could induce differentiation, suggesting a role of RB3 during neuroblastoma differentiation. Moreover, increased RB3 is a unique response protein in the stathmin-related proteins following optical nerve axotomy and RB3 may be a critical target for neurodegenerative diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STMN4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene STMN4
Gene Accession No. P63042, P63043, Q9H169
Gene Alias Lagl; R75359; Rb3; stathmin 4; stathmin-4; stathmin-like 4; stathmin-like protein B3; stathmin-like protein RB3; stathmin-like-protein RB3; Stmn4
Gene Symbols STMN4
Host Species Rabbit
Immunogen A synthesized peptide derived from human STMN4(Accession Q9H169), corresponding to amino acid residues M50-A100.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56471, 79423, 81551
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.