Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STMND1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566968
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566968 Supplier Invitrogen™ Supplier No. PA566968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AEVAFAKGLQR VRSAGFEPSD LQGGKPLKRK KSKCDATLID RNESDESFGV VESDMSYNQA Highest antigen sequence identity to the following orthologs - mouse 51%, rat 49%.

STMND1 gene ontology annotations related to this gene include microtubule depolymerization; neuron projection development; regulation of cytoskeleton organization; regulation of microtubule polymerization or depolymerization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STMND1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STMND1
Gene Accession No. H3BQB6
Gene Alias stathmin domain containing 1; stathmin domain-containing protein 1; STMND1
Gene Symbols STMND1
Host Species Rabbit
Immunogen Recombinant Human STMND1. Recombinant protein control fragment (Product #RP-107612).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 401236
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.