Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ STO-609 acetate
GSA_VAGREENER_CHOICESDP

Catalog No. 155110 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-511-0 10 mg
15-515-0 50 mg
15-511-R 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 15-511-0 Supplier Tocris Bioscience™ Supplier No. 1551/10
Add to Cart
Add to Cart
This item is not returnable. View return policy

Selective CaM kinase kinase inhibitor

STO-609 acetate is a selective, cell-permeable inhibitor of Ca2+-calmodulin-dependent protein kinase kinase (Ki values are 80 and 15 ng/ml for inhibition of CaM-KKα and CaM-KKβ respectively); competes for the ATP-binding site. Displays > 80-fold selectivity over CaMK1, CaMK2, CaMK4, MLCK, PKC, PKA and p42 MAPK.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Chemical Name or Material 7-Oxo-7H-benzimidazo[2,1-a]benz[de]isoquinoline-3-carboxylic acid acetate
CAS 1173022-21-3
Assay Percent Range >98%
Quantity 10 mg
Solubility Soluble to 10mM with sonication in DMSO
Molecular Formula C19H10N2O3.C2H4O2
Formula Weight Observed MW: 374.35

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.