Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoddard Solvent

Catalog No. p-193583
Click to view available options
Quantity:
1 kg
5 kg

CAS: 8052-41-3 | C8H15BrO2 | 223.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8052-41-3
Molecular Formula C8H15BrO2
Molecular Weight (g/mol) 223.11
MDL Number MFCD00132767
InChI Key IMSPZLCTPSXORJ-UHFFFAOYSA-N
Synonym 6-bromo-1-hexanol acetate, 1-acetoxy-6-bromohexane, stoddard solvent, 1-hexanol,6-bromo-,acetate, neo-clear r xylene substitute, w/w benzene, acetic acid 6-bromohexyl ester, 6-acetoxy-1-bromohexane
PubChem CID 11806291
IUPAC Name 6-bromohexyl acetate
SMILES CC(=O)OCCCCCCBr

Specifications

Melting Point -40.0°C
Density 0.7870g/mL
Boiling Point 149.0°C to 213.0°C
Quantity 1 kg
Flash Point >38°C
Packaging Glass Bottle
Physical Form Solvent
Specific Gravity 0.787
UN Number 1268
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Stoddard solvent
Show More Show Less
Hazard Category Danger
Hazard Statement GHS P Statement May be fatal if swallowed and enters airways. Toxic to aquatic life with long lasting effects. Flammable liquid and vapor. Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not breathe dust/fume/gas/mist/vapors/spray. Do not get in eyes, on skin, or on clothing. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do NOT induce vomiting.
EINECSNumber 232-489-3
RTECSNumber WJ8925000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.