Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Stoichiometry of Copper and Silver Nitrate
GREENER_CHOICESCI_ED

Catalog No. S280001 Shop All Aldon Products
Change view
Click to view available options
Class Size:
15 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S280001 15 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S280001 Supplier Innovating Science™ Supplier No. IS2527
Add to Cart
Add to Cart

Introduce students to stoichiometry and mole ratios with the reaction of copper and silver nitrate!

  • Students will watch as beautiful silver crystals are formed on the copper wire, then determine the moles of silver formed and moles of copper reacted to calculate the mole ratio.
  • Using this data, they will write the balanced chemical equation for the reaction, and determine the limiting reactant and their percent yield.
  • Kit contains enough materials for 15 groups of students.
  • Teacher's Manual and Student Study Guide Copymasters are included.

Specifications

Certifications/Compliance Next Generation Science Standards (NGSS)
Color White
Grade 7 to 14
Age 13 to 20
For Use With (Application) Chemistry
Includes 15 Pieces Copper Wire, 10 g Silver Nitrate, 200 mL Acetone
Class Size 15 Lab Groups
Product Type Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.