Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STOM Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5110443
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5110443 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5110443 Supplier Invitrogen™ Supplier No. PA5110443
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KIIKEYERAI IFRLGRILQG GAKGPGLFFI LPCTDSFIKV DMRTISFDIP PQEILTKDSV TISVDGVVYY RVQNATLAVA NITNADSATR LLAQTTLRNV LGTKNLSQIL SDREEIAHNM QSTLDDATDA WGIKVERVEI KDVKLPVQLQ RAMAAEAEAS REARAKVIAA EGEMNASRAL KEASMVITES PAALQLRYLQ TLTTIAAEKN STIVFPLPID MLQGIIGAKH SHLG.

Thought to regulate cation conductance. May regulate ACCN1 and ACCN3 gating.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STOM
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.54 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene STOM
Gene Accession No. P27105, P54116
Gene Alias BND7; EPB7; Epb7.2; EPB72; erythrocyte band 7 integral membrane protein; erythrocyte membrane protein band 7.2 (stomatin); erythrocyte protein band 7.2; erythrocyte protein band 7.2; protein 7.2b; erythrocyte surface protein band 7.2; protein 7.2b; STOM; Stomatin
Gene Symbols STOM
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 55-288 of human STOM (NP_004090.4).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 13830, 2040, 296655
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.