Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STOM Rabbit anti-Human, Polyclonal, Proteintech
SDP

Catalog No. 501727672 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-172-7672 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-172-7672 Supplier Proteintech Group Inc Supplier No. 120461AP150UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Stomatin is an integral membrane protein found in lipid/protein-rich microdomains of almost all human tissues. It was named after the rare human disease haemolytic anaemia hereditary stomatocytosis. Stomatin is implicated in signal transduction and cell communication, and it may regulate cation movement through ion channels and transporters. Absence of Stomatin may cause Na+ and K+ ions to leak into and from erythrocytes. A second function of Stomatin may be to act as a cytoskeletal anchor. Stomatin is a major lipid-raft component of erythrocytes and epithelial cells, and is also an abundant platelet protein. It contains a single hydrophobic domain, close to the N-terminus, and a phosphorylation site.

Specifications

Antigen STOM
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene STOM
Gene Accession No. P27105
Gene Alias BND7, EPB7, EPB72, Protein 7.2b, STOM, stomatin
Gene Symbols STOM
Host Species Rabbit
Immunogen STOM Fusion Protein Ag2676
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2040
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.