Learn More
Novus Biologicals™ Stomach Matched Tumor and Normal Tissue Lysate
Shop All Novus Biologicals Products

Description
Specifications
Specifications
| Tissue | Stomach |
| Quantity | 2 Vials |
| Host Species | Human |
| Content And Storage | Store at −80°C. Avoid freeze-thaw cycles. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.