Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STOML1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559953
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559953 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559953 Supplier Invitrogen™ Supplier No. PA559953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GPGMVLLLPF IDSFQRVDLR TRAFNVPPCK LASKDGAVLS VGADVQFRIW DPVLSVMTVK DLNTATRMTA QNAMTKALLK RPLREIQMEK LKISDQLLLE INDVTRA Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 94%.

Stomatin-like protein 1 (STOML1) is encoded by the STOML1 gene located on chromosome 17 in humans. It is part of a family of five mammalian genes that encode peripheral membrane proteins involved in modulating ion channel functions. STOML1 is known to interact specifically with members of the acid-sensing ion channel (ASIC) family, influencing their proton sensitivity. These interactions help regulate ion transport and signaling pathways, which are critical for cellular homeostasis. In addition, STOML1 has been studied in the context of oral squamous cell carcinoma (OSCC), where it is often overexpressed along with STOML2, potentially affecting cancer progression. Structurally, STOML1 contains a stomatin domain which is crucial for its role in membrane association and ion channel modulation. Its expression in olfactory sensory neurons (OSNs) is comparatively lower than other family members, but it still contributes to the localization and functional integrity of related proteins in these cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STOML1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene STOML1
Gene Accession No. Q9UBI4
Gene Alias 1810015E19Rik; AI847431; EPB72-like 1; EPB72-like protein 1; hUNC-24; MSTP019; Protein unc-24 homolog; RGD1559463; SLP1; SLP-1; stomatin (EBP72)-like 1; stomatin (EPB72)-like 1; stomatin like 1; stomatin-like 1; stomatin-like protein 1; Stomatin-related protein; STOML1; STORP; UNC24; WPB72
Gene Symbols STOML1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STOML1. Recombinant protein control fragment (Product #RP-89968).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9399
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.