Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ STOML3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584854
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584854 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584854 Supplier Invitrogen™ Supplier No. PA584854
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TIVFPLPMNI LEGIGGVSYD NHKKLPNKA Highest antigen sequence indentity to the following orthologs: Rat - 83%, Mouse - 79%.

STOML3 is required for the function of many mechanoreceptors. It modulates mechanotransduction channels and acid-sensing ion channels (ASIC) proteins. The protein potentiates PIEZO1 and PIEZO2 function by increasing their sensitivity to mechanical stimulations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen STOML3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Stoml3
Gene Accession No. Q8TAV4
Gene Alias Epb7.2l; EPB72-like 3; erythrocyte band 7 integral membrane protein; protein 7.2B; SLP3; SLP-3; sro; stomatin (Epb7.2)-like 3; stomatin (EPB72)-like 3; stomatin like 3; stomatin related olfactory protein SRO; stomatin-like protein 3; Stomatin-related olfactory protein; STOML3
Gene Symbols Stoml3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human STOML3. Recombinant protein control fragment (Product #RP-108251).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 161003
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.