Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ STOML3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113228
Description
STOML3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
STOML3 is required for the function of many mechanoreceptors. It modulates mechanotransduction channels and acid-sensing ion channels (ASIC) proteins. The protein potentiates PIEZO1 and PIEZO2 function by increasing their sensitivity to mechanical stimulations.
Specifications
| STOML3 | |
| Polyclonal | |
| Unconjugated | |
| Stoml3 | |
| Epb7.2l; EPB72-like 3; erythrocyte band 7 integral membrane protein; protein 7.2B; SLP3; SLP-3; sro; stomatin (Epb7.2)-like 3; stomatin (EPB72)-like 3; stomatin like 3; stomatin related olfactory protein SRO; stomatin-like protein 3; Stomatin-related olfactory protein; STOML3 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 161003, 229277, 295041 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q6PE84, Q8TAV4 | |
| Stoml3 | |
| Recombinant Human Stomatin-like protein 3 protein (205-291AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction