Learn More
Cytiva™ Stop Plug, 5/16 in. Male
Catalog No. 01257015 Shop All Cytiva Products
Change view
Click to view available options
Packaging Quantity:
5 Fittings
Catalog No. Packaging Quantity
01-257-015 5 Fittings
1 options
This item is not returnable. View return policy
Catalog No. 01-257-015 Supplier Cytiva™ Supplier No. 18111250
NLSU: Get 1 Consumable Product When You Buy 1
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with the ÄKTA™ pure 150 and ÄKTA™ avant 150.
  • Use with ÄKTA pure 150 and ÄKTA avant 150 systems
TRUSTED_SUSTAINABILITY
Specifications
Type Stop Plug
Packaging Quantity 5 Fittings
Thread Size 5/16 in. (m)
Material PEEK