Learn More
Thermo Scientific™ Storage Canes
GSA_VAGREENER_CHOICE
Catalog No. 12565181
Change view
Click to view available options
Type:
Cryo Cane Coder
Cryo Storage Cane
Design:
Coder
Standard
Stepped
Length (Metric):
29 cm
30 cm
Not Applicable
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Design Length (Metric)
12-565-181 Cryo Storage Cane Standard 29 cm
15-350-20 Cryo Storage Cane Stepped 30 cm
15-350-21 Cryo Storage Cane Standard 30 cm
15-350-35 Cryo Cane Coder Coder Not Applicable
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 12-565-181 Supplier Thermo Scientific™ Supplier No. 378441
Add to Cart
Add to Cart
Store various cryostorage tubes in these storage canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
Store Thermo Scientific™ Nalgene™ or Thermo Scientifc™ Nunc™ cryostorage tubes in Thermo Scientific™ Storage Canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
  • Able to store tubes protected with Thermo Scientific™ CryoFlex™ tubing
  • Compatible with color coders that attach to the end of the cane and can be written on for quick identification and inventory management
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
TRUSTED_SUSTAINABILITY
Specifications
Design Standard
Material Aluminum
Type Cryo Storage Cane
Quantity 1/Pk., 50/Cs.
Length (English) 11.31 in.
Length (Metric) 29 cm