Learn More
Thermo Scientific™ Storage Canes



Description
Store Thermo Scientific™ Nalgene™ or Thermo Scientifc™ Nunc™ cryostorage tubes in Thermo Scientific™ Storage Canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
- Able to store tubes protected with Thermo Scientific™ CryoFlex™ tubing
- Compatible with color coders that attach to the end of the cane and can be written on for quick identification and inventory management
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Design | Stepped |
| Material | Aluminum |
| Type | Cryo Storage Cane |
| Quantity | 12/Pk., 48/Cs. |
| Length (English) | 11.31 in. |
| Length (Metric) | 30 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.