Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synthware™ Storage Vessel with High Vacuum Valve
GREENER_CHOICESDP

Catalog No. p-7180042
Click to view available options
Capacity (Metric):
50 mL
100 mL
350 mL

Storage Vessel features a #15 O-ring joint on the sidearm and a high vacuum valve plug to control gas and/or liquid flow into the vessel

Specifications

Color Clear
Capacity (Metric) 50 mL
Material Borosilicate Glass
For Use With (Application) Organic Synthesis
Height (Metric) 190 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.